missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Copper foil, alloy 400, 1.6mm (0.062 in.) thick, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 045231.HB
This item is not returnable.
View return policy
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Nickel Copper foil, Alloy 400 | |
Ni:Cu; 67:33 wt% | |
300 x 300 mm |
1.6mm (0.062 in.) thick | |
MFCD00801098 | |
Foil |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nickel Copper foil, alloy 400, 1.6mm (0.062 in.) thick, Thermo Scientific™ > 300x300mm
Spot an opportunity for improvement?Share a Content Correction