missing translation for 'onlineSavingsMsg'
Learn More

Nickel Copper foil, alloy 400, 1.6mm (0.062 in.) thick, Thermo Scientific™

£181.91 - £453.00

Specifications

Chemical Name or Material Nickel Copper foil, Alloy 400
Name Note 1.6mm (0.062 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cu; 67:33 wt%
MDL Number MFCD00801098
Form Foil
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15416845
View Documents
Thermo Scientific Alfa Aesar
045231.GT
150 x 150 mm
£181.91
Each
Please to purchase this item. Need a web account? Register with us today!
15426845
View Documents
Thermo Scientific Alfa Aesar
045231.HB
300 x 300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Nickel Copper foil, Alloy 400
Ni:Cu; 67:33 wt%
Foil
1.6mm (0.062 in.) thick
MFCD00801098
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel Copper foil, alloy 400, 1.6mm (0.062 in.) thick, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.