missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Copper Iron Manganese rod, 1.27cm (0.5 in.) dia., Thermo Scientific™
£85.90 - £317.00
Specifications
Chemical Name or Material | Nickel Copper Iron Manganese rod |
---|---|
Name Note | 1.27cm (0.5 in.) dia., cold drawn stress relieved, ASTM B164 |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cu:Fe:Mn; 63:28 to 34:2.5 max:2 max wt% |
MDL Number | MFCD02091727 |
Weight | ≈104g/10cm |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nickel Copper Iron Manganese rod | |
Ni:Cu:Fe:Mn; 63:28 to 34:2.5 max:2 max wt% | |
≈104g/10cm |
1.27cm (0.5 in.) dia., cold drawn stress relieved, ASTM B164 | |
MFCD02091727 | |
Rod |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Nickel Copper Iron Manganese rod
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
- Carcinogenicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .