missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Nickel Crucible

Nickel Electro™ Nickel Crucible is crevice free and suitable for temperatures up to 350°C.

Brand:  Nickel Electro™ 39

 View more versions of this product

Product Code. 11429599

  • £101.00 / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 99.6% nickel purity
  • Crevice free
  • High thermal and electrical conductivities
  • Low gas content
  • Low vapor pressure
  • Cost effective replacement for platinum
Specifications

Specifications

Nickel
2 mm
Autoclavable
50 mm
Each
70 mL
50 mm
Crucible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel Electro™ Nickel Crucible >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.