Learn More
Nickel(II) bromide trihydrate, 98%, Thermo Scientific Chemicals
£76.28 - £368.84
Chemical Identifiers
CAS | 7789-49-3 |
---|---|
Molecular Formula | Br2H6NiO3 |
Molecular Weight (g/mol) | 272.546 |
MDL Number | MFCD00011141 |
InChI Key | UQPSGBZICXWIAG-UHFFFAOYSA-L |
Synonym | nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate at |
PubChem CID | 53471882 |
IUPAC Name | dibromonickel;trihydrate |
SMILES | O.O.O.[Ni](Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353758
|
Thermo Scientific Alfa Aesar
040242.18 |
50 g |
£76.28
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343758
|
Thermo Scientific Alfa Aesar
040242.30 |
250 g |
£368.84
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in Soporific and sedatives pharmaceutical Industry, Photosensitive industry, Analytical Chemistry, and also electrolyte in high energy battery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-49-3 | |
272.546 | |
UQPSGBZICXWIAG-UHFFFAOYSA-L | |
53471882 | |
O.O.O.[Ni](Br)Br |
Br2H6NiO3 | |
MFCD00011141 | |
nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate at | |
dibromonickel;trihydrate |
Specifications
300°C (in -3H2O) | |
Crystalline Granules | |
MFCD00011141 | |
14,6502 | |
Soluble in water, alcohol | |
O.O.O.[Ni](Br)Br | |
272.546 | |
272.58 (218.53 Anhydrous) | |
Odorless | |
Nickel(II) bromide trihydrate |
7789-49-3 | |
Br2H6NiO3 | |
UN3077 | |
nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate at | |
UQPSGBZICXWIAG-UHFFFAOYSA-L | |
dibromonickel;trihydrate | |
53471882 | |
98% | |
Hygroscopic |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H341-H350-H360-H372
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 236-665-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nickel(II) bromide trihydrate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1A
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P363-Wash contaminated clothing before reuse.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Nickel(II) bromide trihydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.