Learn More
Nickel(II) carbonate hydroxide hydrate, for analysis, (ca 45-47% Ni), Thermo Scientific Chemicals
£26.70 - £91.30
Chemical Identifiers
CAS | 39430-27-8, 65405-96-1 |
---|---|
Molecular Formula | CH4Ni3O7·xH2O |
Molecular Weight (g/mol) | 304.11 |
MDL Number | MFCD00150262 |
Synonym | Basic nickel carbonate |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39430-27-8, 65405-96-1 | |
304.11 | |
Basic nickel carbonate |
CH4Ni3O7·xH2O | |
MFCD00150262 |
Specifications
39430-27-8, 65405-96-1 | |
Powder | |
2Ni(OH)2·xH2O | |
13, 6527 | |
Solubility in water: insoluble. Other solubilities: soluble in ammonia and in diluted acids | |
304.11 | |
Analytical |
Green | |
CH4Ni3O7·xH2O | |
MFCD00150262 | |
Basic nickel carbonate | |
304.11 | |
≥45% (NI) | |
Nickel(II) carbonate hydroxide hydrate, ca 45-47% Ni, For analysis |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause cancer by inhalation.
Suspected of causing genetic defects
GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.