missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

£121.61 - £478.38

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11315747
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043441.06
5 g
£121.61
5g
Please to purchase this item. Need a web account? Register with us today!
11325747
View Documents
Thermo Scientific Alfa Aesar
043441.14
25 g
£478.38
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel(II) chloride is used in electroplating, electroforming ,electroless plating, tea and coffee plantation, dye mordant. It is also used in semiconducting metal-containing polymer synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7718-54-9
129.593
QMMRZOWCJAIUJA-UHFFFAOYSA-L
CHEBI:34887
Cl[Ni]Cl
Cl2Ni
MFCD00011142
24385
dichloronickel

Specifications

1,001°C
Yellow
Cl2Ni
MFCD00011142
14,6505
QMMRZOWCJAIUJA-UHFFFAOYSA-L
dichloronickel
24385
129.6
Odorless
3.55 g/mL
Nickel (II) chloride, Anhydrous
7718-54-9
Crystalline Powder
NiCl2
UN3288
It is soluble in water,ethylene glycol,ethanol, ammonium hydroxide. It is insoluble in ammonia, nitric acid.
Cl[Ni]Cl
129.593
CHEBI:34887
99.99%
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H334-H350-H360-H372-H341-H315-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c

H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-743-0

missing translation for 'rtecsNumber' : QR6475000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only