missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) formate dihydrate, Thermo Scientific Chemicals Product Code.: 11321349

Nickel(II) formate dihydrate, Thermo Scientific Chemicals

Product Code. 11321349
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11321349

Brand: Thermo Scientific Alfa Aesar 013801.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Nickel Catalyst Preparations

Nickel(II) formate dihydrate is used in nickel catalyst preparations. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15694-70-9
Molecular Formula Ni(HCO2)2·2H2O
MDL Number MFCD00153034

Specifications

Melting Point 130°C to 140°C-2H2O
Color Green
Physical Form Powder
Quantity 100 g
Solubility Information In the range 180-200° decomposes to Ni,CO,CO2,H2,H2O,CH4. Partially soluble in water
Formula Weight 184.77 (148.74 Anhydrous)
Appearance Green powder
Decomposition Information In the range 180° to 200°C decomposes to Ni,CO,CO2
Density 2.15 g/cm3
Chemical Name or Material Nickel(II) formate dihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX12G033 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number013801.22,013801.36,013801.A3
Lot NumberP31G024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013801.22,013801.36,013801.A3
Lot NumberD30T033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013801.22,013801.36,013801.A3
Lot NumberC22L24 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013801.22,013801.36,013801.A3
Lot NumberS30G057 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013801.22,013801.36,013801.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) formate dihydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.