missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals Product Code.: 11428740

Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11428740
2.5 kg
Click to view available options
Quantity:
100 g
500 g
1000 g
2.5 kg
5000 g
This item is not returnable. View return policy

Product Code. 11428740

Brand: Thermo Scientific Alfa Aesar A15540.A4

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

Solubility
Very soluble in water. Soluble in ethanol.

Notes
As an oxidizer, it is incompatible with reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6Show MoreShow Less
PubChem CID 61630
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 137°C
Melting Point 56.7°C
Quantity 2.5 kg
Linear Formula Ni(NO3)2·6H2O
UN Number UN2725
Merck Index 14,6513
Solubility Information Very soluble in water. Soluble in ethanol.
IUPAC Name nickel(2+) hexahydrate dinitrate
Formula Weight 290.80 (182.70 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.05
Chemical Name or Material Nickel(II) nitrate hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247755 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA15540.0I,A15540.22,A15540.0B,A15540.36,A15540.A4
Lot Number10241541 Certificate TypeCertificate of Analysis Date23/06/2022 Catalog NumberA15540.0I,A15540.22,A15540.0B,A15540.36,A15540.A4
Lot Number10238945 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15540.0I,A15540.22,A15540.0B,A15540.36,A15540.A4
Lot Number10145685 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15540.0I,A15540.22,A15540.0B,A15540.36,A15540.A4
Lot Number10220047 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15540.0I,A15540.22,A15540.0B,A15540.36,A15540.A4
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Oxidising solid Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : Nickel(II) nitrate,hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.