Learn More
Nickel(II) nitrate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Boiling Point | 137.0°C |
Melting Point | 56.7°C |
Color | Blue to Green |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Physical Form | Crystalline Powder |
Quantity | 10 kg |
Assay Percent Range | 98.5% min. (Complexometry) |
Linear Formula | Ni(NO3)2·6H2O |
Merck Index | 15,6598 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463184 | Certificate of Analysis | 12/06/2024 | 223155000,223150100,223150025 |
A0457742 | Certificate of Analysis | 14/11/2023 | 223155000,223150100,223150025 |
A0449417 | Certificate of Analysis | 07/02/2023 | 223155000,223150100,223150025 |
A0395507 | Certificate of Analysis | 16/12/2022 | 223155000,223150100,223150025 |
A0410823 | Certificate of Analysis | 13/12/2022 | 223155000,223150100,223150025 |
Safety and Handling





- Nickel(II) nitrate hexahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.