missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) oxide, black, 76% Ni, Thermo Scientific Chemicals Product Code.: 10318550

Nickel(II) oxide, black, 76% Ni, Thermo Scientific Chemicals

Product Code. 10318550
100 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10318550

Brand: Thermo Scientific Acros 223121000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxideShow MoreShow Less
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Color Black-Gray
Physical Form Powder
Quantity 100 g
Linear Formula NiO
Merck Index 15, 6597
Formula Weight 74.7
Percent Purity 75 to 77% (Ni)
Packaging Glass bottle
Density 6.67g/mL
Chemical Name or Material Nickel(II) oxide, black, 76% Ni

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467712 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number223125000,223121000,223120025
Lot NumberA0460763 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number223125000,223121000,223120025
Lot NumberA0402753 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number223125000,223120050,223121000,223120025
Lot NumberA0448323 Certificate TypeCertificate of Analysis Date06/12/2022 Catalog Number223125000,223120050,223121000,223120025
Lot NumberA0446314 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog Number223125000,223120050,223121000,223120025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) oxide, black, 76% Ni, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.