Learn More
Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3}), Thermo Scientific Chemicals
Description
Nickel(II) oxide is used in fuel cell electrodes, storage batteries, electronic circuit boards, electrical ceramics, and as a pigment for ceramics and glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1984°C |
Color | Black |
Physical Form | Powder |
Quantity | 1 kg |
Merck Index | 14,6512 |
Solubility Information | Soluble in acids. Insoluble in water. |
Formula Weight | 74.71 |
Concentration or Composition (by Analyte or Components) | Ni 76%, (contains Ni2O3) |
Odor | Odorless |
Density | 6.67 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
62500085 | Certificate of Analysis | 14/02/2025 | 087302.30,087302.A1 |
S11G025 | Certificate of Analysis | 02/04/2024 | 087302.30,087302.A1 |
Y01J010 | Certificate of Analysis | 07/11/2023 | 087302.30,087302.A1 |
N07I048 | Certificate of Analysis | 12/04/2022 | 087302.30,087302.A1 |
C18U023 | Certificate of Analysis | 08/12/2021 | 087302.30,087302.A1 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.