missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 53.0°C |
Color | Blue-Green |
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Physical Form | Crystals |
Quantity | 500 g |
Assay Percent Range | 98.0 to 102% (Complexometry) |
Linear Formula | NiSO4·6H2O |
Merck Index | 15,6602 |
Solubility Information | Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0447702 | Certificate of Analysis | 31/01/2025 | 415615000,415610051,415611000,415610010 |
A0464973 | Certificate of Analysis | 12/08/2024 | 415615000,415610051,415610010,415611000 |
A0460556 | Certificate of Analysis | 14/03/2024 | 415615000,415610051,415610010,415611000 |
A0457952 | Certificate of Analysis | 30/11/2023 | 415615000,415610051,415610010,415611000 |
A0447702 | Certificate of Analysis | 28/11/2023 | 415615000,415610051,415610010,415611000 |
Safety and Handling



Product Identifier
- Nickel(II) sulfate hexahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction