Learn More
Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals
Form: approx. 40.1g/150 x 300mm
Brand: Thermo Scientific Alfa Aesar 045092.@O
Code : NEW
Additional Details : Weight : 0.01000kg
Description
It is most commonly used as a high temperature, corrosion resistant metal alloy in steel, cast iron and super alloys for the military and defense industry, semiconductor industry and specialty machine shops.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Iron Molybdenum foil | |
Ni:Fe:Mo; 80:15:4.2 wt% | |
MFCD09910537 | |
Insoluble aq. soln.. | |
Foil |
0.102 mm (0.004 in.) thick, Annealed | |
Ni:Fe:Mo | |
300 x 600 mm | |
∼40.1 g/150 x 300 mm |
Safety and Handling
- Nickel Iron Molybdenum foil
Signal Word
- Danger
Hazard Category
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Nickel
RUO – Research Use Only