missing translation for 'onlineSavingsMsg'
Learn More

Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals

Form: approx. 40.1g/150 x 300mm

£46.60 - £101.00

Specifications

Chemical Name or Material Nickel Iron Molybdenum foil
Avg. Mol. Wt. or Mol. Wt. Range Ni:Fe:Mo; 80:15:4.2 wt%
Molecular Formula Ni:Fe:Mo
MDL Number MFCD09910537
Form Foil
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15436735
View Documents
Thermo Scientific Alfa Aesar
045092.4K
150 x 300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15456735
View Documents
Thermo Scientific Alfa Aesar
045092.HB
300 x 300 mm
£63.82
Each
Please to purchase this item. Need a web account? Register with us today!
15426735
View Documents
Thermo Scientific Alfa Aesar
045092.@O
300 x 600 mm
£101.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is most commonly used as a high temperature, corrosion resistant metal alloy in steel, cast iron and super alloys for the military and defense industry, semiconductor industry and specialty machine shops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel Iron Molybdenum foil
Ni:Fe:Mo
Foil
Ni:Fe:Mo; 80:15:4.2 wt%
MFCD09910537
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350-H372

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only