missing translation for 'onlineSavingsMsg'
Learn More

Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals

Form: approx. 40.1g/150 x 300mm

Brand:  Thermo Scientific Alfa Aesar 045092.HB

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15456735

  • £62.90 / Each
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It is most commonly used as a high temperature, corrosion resistant metal alloy in steel, cast iron and super alloys for the military and defense industry, semiconductor industry and specialty machine shops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel Iron Molybdenum foil
Ni:Fe:Mo; 80:15:4.2 wt%
MFCD09910537
Insoluble in water.
Foil
0.102mm (0.004 in.) thick, Annealed
Ni:Fe:Mo
300 x 300 mm
≈40.1g/150x300mm
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only