missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&L Enterprises™ Hoffman Clips
An excellent option for sealing flexible tubing in the laboratory. R&L Enterprises™ Hoffman Clips are made of nickel plated brass to prevent corrosion.
£179.00 - £191.48
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
12823183
|
R&L Enterprises™
9053 |
For use with 30mm tubing |
£179.00
Pack of 10 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
12764396
|
R&L Enterprises™
9054 |
For use with 40mm O.D. tubing |
£191.48
Pack of 10 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Description
- Either side or bottom opening
- With clamping screw hinges
Specifications
Nickel-plated | |
10 Pack | |
Clip |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&L Enterprises™ Hoffman Clips