Learn More
R&L Enterprises™ Hoffman Clips
An excellent option for sealing flexible tubing in the laboratory. R&L Enterprises™ Hoffman Clips are made of nickel plated brass to prevent corrosion.
Brand:  R&L Enterprises™ 9053
 View more versions of this product
Product Code. 12823183
  • £179.00 / Pack of 10
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Either side or bottom opening
  • Available for a range of tube thicknesses
  • Threaded bar with thumbscrew
Specifications
Specifications
Nickel-plated
For use with 30mm tubing
10 Pack
Clip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Hoffman Clips > For Use With: For use with 30mm tubing