Learn More
Thermo Scientific™ Oxoid™ C-N Supplement
Oxoid C-N Supplement is used for the isolation of Pseudomonas aeruginosa.
Brand: Thermo Scientific™ SR0102E
38.98 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Oxoid C-N Supplement is used for the isolation of Pseudomonas aeruginosa.
- Add to to Pseudomonas Agar Base, Part No. CM0559B
- Each vial supplements 500mL of medium

Specifications
C-N Supplement | |
Microbiology Media | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6226620 | Certificate of Analysis | 17/03/2025 | SR0102E |
6199408 | Certificate of Analysis | 16/12/2024 | SR0102E |
6180718 | Certificate of Analysis | 04/11/2024 | SR0102E |
6150832 | Certificate of Analysis | 22/08/2024 | SR0102E |
6113179 | Certificate of Analysis | 18/06/2024 | SR0102E |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
83Hours
17Minutes
27Seconds
6New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.