missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pseudomonas Agar Base (Dehydrated)

Selectively isolate Pseudomonasspecies from a range of samples with this medium.

Brand:  Thermo Scientific™ CM0559B

79.09 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10157992

  • £173.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Selectively isolate Pseudomonas species from a range of samples with Thermo Scientific™ Oxoid™ Pseudomonas Agar Base (Dehydrated). The medium is modification of King's A Medium1 and contains magnesium chloride and potassium sulphate for improved pigment production and recovery of Pseudomonas aeruginosa while strongly suppressing Klebsiella, Proteus and Providencia spp.

Pseudomonas Agar Base is designed so that by the addition of the appropriate supplement (SR0102 or SR0103) the medium becomes selective for Pseudomonas aeruginosa or Pseudomonas spp. generally.

  • Selective: Pseudomonas aeruginosa forms fluorescent green colonies while the growth of Klebsiella, Proteus, and Providencia is strongly suppressed
  • Enhanced pigment production aids identification

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pseudomonas Agar Base
Agar
For 10.3L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224029 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberCM0559B
Lot Number6209834 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog NumberCM0559B
Lot Number6212220 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog NumberCM0559B
Lot Number6205545 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberCM0559B
Lot Number6182807 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberCM0559B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pseudomonas Agar Base (Dehydrated) > Pseudomonas Agar Base; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only