Learn More
Thermo Scientific™ Oxoid™ Phosphate Buffered Saline Tablets
Oxoid Phosphate Buffered Saline Tablets (Dulbecco A) is used for preparation of balanced salt solution without calcium and magnesium.
Brand: Thermo Scientific™ BR0014G
28.79 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Oxoid Phosphate Buffered Saline Tablets (Dulbecco A) is used for preparation of balanced salt solution without calcium and magnesium.

Specifications
Phosphate Buffered Saline Tablets | |
Media Supplement | |
Each tab for 100mL |
Tablet | |
Each |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4476641 | Certificate of Analysis | 17/03/2025 | BR0014G |
4463565 | Certificate of Analysis | 04/10/2024 | BR0014G |
4446725 | Certificate of Analysis | 17/05/2024 | BR0014G |
4428545 | Certificate of Analysis | 09/05/2024 | BR0014G |
4411091 | Certificate of Analysis | 06/01/2024 | BR0014G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only