Learn More
Palintest™ Photometer Tablet Reagents
Photometer reagents in convenient tablet form. Palintest™; Photmoeter Tablet Reagents are available for a variety of water testing needs.
Brand: Palintest™ PM179
Description
- Convenient tablet form.
- Easy to use.
- Available for a variety of water testing needs.
Specifications
50 | |
0 to 1.5mg/L | |
Fluoride Test Kit |
50/Pk. | |
50 Tests |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- X50 TEST KIT/PHOTOM FLUORID 1,5PPM
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash contaminated skin thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see medical advice on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : lithium hydroxide, boric acid, sulphamicacid, zirconium(IV)oxychloride octahydrate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.