Learn More
Epredia™ Histoplast Paraffin
Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.
Brand: Epredia™ 8330
137.25 GBP valid until 2025-04-30
Use promo code "24247" to get your promotional price.
Description
- Made of paraffin and plastic polymers
- Delivers excellent processing and yields thin, wrinkle-free sections
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).

Specifications
56°C | |
Paraffin PE | |
8 x 1 Kg |
Safety and Handling
- Warning
Hazard Category
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H335-May cause respiratory irritation.
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P314-Get medical advice/attention if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : paraffin waxes and hydrocarbon waxes, polyisobutylene, 2,6-ditertbutyl-p-cresol
Special Offers
-
Multiple Manufacturers
Life Sciences Monthly Promotion
Discover exclusive monthly deals on life sciences products to advance your research and innovation.Promo Code: 24247Expires: 2025-04-30
New Offer Save Up to 52%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.