missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin - Agarose
Streptavidin - Agarose
Brand: Molecular Probes™ 434341
This item is not returnable.
View return policy
Description
Reactivity: SA-Seph 4B reacts with biotin.
Specifications
1 mg Streptavidin/mL Sepharose Gel | |
Store in refrigerator (2–8°C). | |
Liquid Suspension |
2 mL | |
Wet Ice | |
Streptavidin Conjugate (Fluorescent) |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Streptavidin - Agarose
Version: 06/09
File size: 22 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
YE367760 | Certificate of Analysis | 26/05/2023 | 434341 |
XB325169 | Certificate of Analysis | 17/05/2022 | 434341 |
WC315875 | Certificate of Analysis | 23/03/2021 | 434341 |
UH286917 | Certificate of Analysis | 13/08/2019 | 434341 |
TJ270004 | Certificate of Analysis | 30/10/2018 | 434341 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molecular Probes™ Streptavidin - Agarose > 2mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction