missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Histoplast Paraffin

Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.

Brand:  Epredia™ 8332

Code : A0

Additional Details : Weight : 8.00000kg


 View more versions of this product

Product Code. 12603036

  • £91.70 / Pack of 8
Estimated Shipment: 29-07-2024
to see stock.
Add to basket

Description

Description

  • Made of paraffin and plastic polymers
  • Delivers excellent processing and yields thin, wrinkle-free sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).


Specifications

Specifications

50° to 54°C
Paraffin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H335-May cause respiratory irritation.
  • H371-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P314-Get medical advice/attention if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : parrafin waxes and hydrocarbon waxes, polyisobutylene, 2,6-di-tert-butyl-p-cresol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers