Learn More
Nicotinic acid, 99.5%, Thermo Scientific Chemicals
£22.71 - £954.75
Chemical Identifiers
CAS | 59-67-6 |
---|---|
Molecular Formula | C6H5NO2 |
Molecular Weight (g/mol) | 123.11 |
MDL Number | MFCD00006391 |
InChI Key | PVNIIMVLHYAWGP-UHFFFAOYSA-N |
Synonym | nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil |
PubChem CID | 938 |
ChEBI | CHEBI:15940 |
IUPAC Name | pyridine-3-carboxylic acid |
SMILES | OC(=O)C1=CC=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10591561
|
Thermo Scientific Acros
128290050 |
5 g | Glass Bottle |
£22.71
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10591751
|
Thermo Scientific Acros
128291000 |
100 g | Plastic bottle |
£23.62
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10081320
|
Thermo Scientific Acros
128295000 |
500 g | Plastic bottle |
£69.85
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10722374
|
Thermo Scientific Acros
128290025 |
2.5 kg | Plastic bottle |
£272.36
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15119173
|
Thermo Scientific Acros
128290100 |
10 kg | Plastic drum |
£954.75
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59-67-6 | |
123.11 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
938 | |
pyridine-3-carboxylic acid |
C6H5NO2 | |
MFCD00006391 | |
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
CHEBI:15940 | |
OC(=O)C1=CC=CN=C1 |
Specifications
59-67-6 | |
100.0 | |
White | |
Authentic | |
C6H5NO2 | |
22,38 | |
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
pyridine-3-carboxylic acid | |
938 | |
123.11 | |
Powder |
99.4 | |
236.0°C to 239.0°C | |
1% max. (105°C, 1 hr.) | |
99.5% | |
MFCD00006391 | |
15,661 | |
Solubility in water: 17g/L (20°C). Other solubilities: slightly soluble in ethanol (10g/L at 20°C),eth,er,soluble in propylene glycol,in alkali hydroxides,and carbonates,freely soluble in boiling water or boiling ethanol | |
OC(=O)C1=CC=CN=C1 | |
123.11 | |
CHEBI:15940 | |
99.5% | |
Nicotinic acid, 99.5% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-441-0
missing translation for 'rtecsNumber' : QT0525000
missing translation for 'tsca' : TSCA
- Nicotinic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.