missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nigrosin, alcohol soluble, Thermo Scientific Chemicals
£14.10 - £59.30
Chemical Identifiers
CAS | 11099-03-9 |
---|---|
Molecular Formula | C8H19ClN2O |
Molecular Weight (g/mol) | 194.703 |
MDL Number | MFCD00149005 |
InChI Key | GBPWTMZRCAESKB-UHFFFAOYSA-N |
Synonym | Solvent Black 5; C.I. 50415 |
PubChem CID | 19828004 |
IUPAC Name | 2-amino-N,N-dipropylacetamide;hydrochloride |
SMILES | CCCN(CCC)C(=O)CN.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15453809
|
Thermo Scientific Alfa Aesar
J61186.14 |
25 g |
£14.10
25g |
In Stock
Log in to see stock. |
|||||
15463809
|
Thermo Scientific Alfa Aesar
J61186.22 |
100 g |
£34.20
100g |
In Stock
Log in to see stock. |
|||||
15473809
|
Thermo Scientific Alfa Aesar
J61186.A1 |
1 kg |
£59.30
1kg |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
11099-03-9 | |
194.703 | |
GBPWTMZRCAESKB-UHFFFAOYSA-N | |
19828004 | |
CCCN(CCC)C(=O)CN.Cl |
C8H19ClN2O | |
MFCD00149005 | |
Solvent Black 5; C.I. 50415 | |
2-amino-N,N-dipropylacetamide;hydrochloride |
Specifications
11099-03-9 | |
MFCD00149005 | |
Soluble in alcohol | |
CCCN(CCC)C(=O)CN.Cl | |
194.703 | |
Black | |
Nigrosin |
C8H19ClN2O | |
Solvent Black 5; C.I. 50415 | |
GBPWTMZRCAESKB-UHFFFAOYSA-N | |
2-amino-N,N-dipropylacetamide;hydrochloride | |
19828004 | |
Powder |
Safety and Handling
missing translation for 'rtecsNumber' : GE5800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only