missing translation for 'onlineSavingsMsg'
Learn More

Niobium(IV) selenide, 99.8% (metals basis), Thermo Scientific Chemicals

£87.54 - £310.55

Chemical Identifiers

CAS 12034-77-4
Molecular Formula NbSe2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00016228
InChI Key CXRFFSKFQFGBOT-UHFFFAOYSA-N
Synonym niobium selenide, columbium selenide, niobium diselenide, diselanylideneniobium, bis selanylidene niobium, niobium diselenide, crystal, niobium diselenide trace metals basis, niobium iv selenide trace metals basis 10gShow MoreShow Less
PubChem CID 82841
IUPAC Name bis(selanylidene)niobium
SMILES [Se]=[Nb]=[Se]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11382188
SDS View Documents
Thermo Scientific Alfa Aesar
013101.09
10 g
£87.54
10g
Please to purchase this item. Need a web account? Register with us today!
11392188
SDS View Documents
Thermo Scientific Alfa Aesar
013101.18
50 g
£310.55
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a potential substance for thin film solar cells as back contact material. It is widely used in electricity brush & special bearings because of its conductivity and lubricity property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

missing translation for 'einecsNumber' : 234-811-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium(IV) selenide, 99.8% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.