missing translation for 'onlineSavingsMsg'
Learn More
Niobium pieces, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11340709

Niobium pieces, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11340709
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11340709

Brand: Thermo Scientific Alfa Aesar 000241.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-03-1
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Assay Percent Range 99.8% (metals basis)
Quantity 100 g
Percent Purity 99.8%
Form Pieces
Assay (metals basis)
Chemical Name or Material Niobium pieces

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ18K003 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number000241.22,000241.36
Lot NumberU15J076 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number000241.22,000241.36
Lot NumberN11C040 Certificate TypeCertificate of Analysis Date26/05/2022 Catalog Number000241.22,000241.36
Lot NumberN23I068 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000241.22,000241.36
Lot NumberB08Q22 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000241.22,000241.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium pieces, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.