missing translation for 'onlineSavingsMsg'
Learn More
Niobium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals Product Code.: 15454815

Niobium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 15454815
1 Ea.
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy

Product Code. 15454815

Brand: Thermo Scientific Alfa Aesar 040882.KS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Niobium sputtering target is used in high temperature engineering products. It is also used as an alloying agent for certain steels where it greatly improves the strength of the resulting material. It is also finds applications in atomic reactors due to its corrosion resistance and, when combined with either tin (Nb3Sn) or zirconium, it has a high degree of superconductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 7440-03-1
Odor Odorless
Molecular Formula Nb
MDL Number MFCD00011126
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
Solubility Information Insoluble in water.
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES [Nb]
IUPAC Name niobium
Molecular Weight (g/mol) 92.91
PubChem CID 23936
ChEBI CHEBI:33344
Percent Purity 99.95%
Assay (metals basis excluding Ta)
Chemical Name or Material Niobium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC07R026 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040882.KS
Lot NumberK16P24 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040882.KS
Lot NumberP15F062 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040882.KS
Lot NumberE12R015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040882.KS
Lot NumberJ16L02 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040882.KS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.