missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals

£65.53 - £223.11

Chemical Identifiers

CAS 10026-12-7
Molecular Formula Cl5Nb
Molecular Weight (g/mol) 270.16
MDL Number MFCD00011127
InChI Key YHBDIEWMOMLKOO-UHFFFAOYSA-I
Synonym niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
PubChem CID 24818
IUPAC Name pentachloroniobium
SMILES Cl[Nb](Cl)(Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15780829
View Documents Promotion Details
Thermo Scientific Acros
201690100
10 g Glass bottle
£65.53
10g
Please to purchase this item. Need a web account? Register with us today!
10398670
View Documents Promotion Details
Thermo Scientific
201690500
50 g Glass bottle
£223.11
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10026-12-7
270.16
YHBDIEWMOMLKOO-UHFFFAOYSA-I
24818
Cl[Nb](Cl)(Cl)(Cl)Cl
Cl5Nb
MFCD00011127
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
pentachloroniobium

Specifications

254.0°C
10026-12-7
99.75
Fine Crystalline Powder
Cl5Nb
MFCD00011127
15, 6643
Solubility in water: reacts with water. Other solubilities: soluble in hcl, soluble in ccl4, soluble in ethanol
Cl[Nb](Cl)(Cl)(Cl)Cl
270.16
270.16
Niobium(V) chloride, 99.8%
204.7°C to 209.5°C
Yellow
100.0
99.75% min
NbCl5
14,214
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
YHBDIEWMOMLKOO-UHFFFAOYSA-I
pentachloroniobium
24818
99.8%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-059-8

missing translation for 'rtecsNumber' : QU0350000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only