Learn More
Niobium(V) oxide, 99.95% (metals basis), Thermo Scientific Chemicals
For Vacuum Deposition
£75.38 - £248.24
Chemical Identifiers
CAS | 1313-96-8 |
---|---|
Molecular Formula | Nb2O5 |
Molecular Weight (g/mol) | 265.81 |
MDL Number | MFCD00011128 |
InChI Key | ZKATWMILCYLAPD-UHFFFAOYSA-N |
Synonym | niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxdeShow More |
PubChem CID | 123105 |
SMILES | O=[Nb](=O)O[Nb](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357938
|
Thermo Scientific Alfa Aesar
036478.14 |
25 g |
£75.38
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347938
|
Thermo Scientific Alfa Aesar
036478.22 |
100 g |
£248.24
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Niobium(V) oxide is used for vacuum deposition, as an alternate electrode to lithium metal in advanced fuel cells, Applied in the solid state formation of an unusual cation deficient perovskite. And it is also used in Electrical and Electronic Products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-96-8 | |
265.81 | |
ZKATWMILCYLAPD-UHFFFAOYSA-N | |
123105 |
Nb2O5 | |
MFCD00011128 | |
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxdeShow More | |
O=[Nb](=O)O[Nb](=O)=O |
Specifications
1520°C | |
White | |
99.95% (metals basis) | |
MFCD00011128 | |
Soluble in HF. Insoluble in water. | |
O=[Nb](=O)O[Nb](=O)=O | |
265.81 | |
265.81 | |
White 3-12mm Sintered Lump | |
Niobium(V) oxide |
1313-96-8 | |
Sintered Lump | |
Nb2O5 | |
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde | |
ZKATWMILCYLAPD-UHFFFAOYSA-N | |
[(dioxoniobio)oxy]dioxoniobium | |
123105 | |
Odorless | |
4.6 |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'einecsNumber' : 215-213-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.