Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
Offers the highest possible pH precision. Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets are specified to three decimal places and come with a printed certificate that guarantees traceability. Individual packets guarantee fresh solution for every calibration, as well as maximum ease of use.
Brand: Mettler Toledo™ 30111137
127.11 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 7732-18-5 Weight : 0.60000kg
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
- NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
- Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
- A test certificate and detailed label with a temperature table comes with every box
- All SDS and labels contain information according to GHS (Globally Harmonized System)
Specifications
NIST/DIN Buffer pH 6.865 sachets 30x20mL | |
pH measurement, electrode calibration | |
Sachet | |
30 x 20 mL |
pH Electrode | |
Yes | |
6.865 (at 25°C) |