Learn More
Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals
Description
Nitric acid, 65-70% is used in as a nitrating or an oxidizing agent in organic synthesis. It is industrially used for the manufacturing of fertilizers. Further, it finds use in manufacture of explosives, nylon precursors, specialty organic compounds and drug intermediates.It is also used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple. It is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless to Yellow |
Boiling Point | 121°C |
Concentration | 65 to 70% |
Density | 1.41 g/mL |
Odor | Pungent |
Quantity | 500 mL |
Physical Form | Liquid |
UN Number | UN2031 |
Merck Index | 14,6577 |
Solubility Information | Miscible with water. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
62500061 | Certificate of Analysis | 05/02/2025 | 010984.AC,010984.AK,010984.AP |
62400561 | Certificate of Analysis | 28/01/2025 | 010984.AC,010984.AK,010984.AP |
S22J030 | Certificate of Analysis | 28/06/2023 | 010984.AC,010984,010984.AK,010984.AP |
S05J035 | Certificate of Analysis | 13/06/2023 | 010984.AP |
Q19J028 | Certificate of Analysis | 25/04/2023 | 010984.AP |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
55Hours
9Minutes
42Seconds
8New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.