missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
HNO3, CAS Number-7697-37-2, acide nitrique, azotic acid, hydrogen nitrate, salpetersaeure, rfna, nitryl hydroxide, nitrous fumes, nital, acidum nitricum, aqua fortis, 2.5L, 122 deg.C, CHEBI:48107, Colorless Yellow, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -42 deg.C, 11349, 63.012
£36.25 - £94.10
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10543674
|
Fisher Chemical
N/2300/PB08 |
500 mL | Plastic Coated Glass bottle |
£36.25
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10634732
|
Fisher Chemical
N/2300/PB15 |
1 L | HDPE plastic bottle |
£39.60
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10654732
|
Fisher Chemical
N/2300/PB17 |
2.5 L | HDPE plastic bottle |
£70.30
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378433
|
Fisher Chemical
N/2300/PC17 |
2.5 L | Plastic Coated Glass bottle |
£94.10
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HNO3, CAS Number-7697-37-2, acide nitrique, azotic acid, hydrogen nitrate, salpetersaeure, rfna, nitryl hydroxide, nitrous fumes, nital, acidum nitricum, aqua fortis, 2.5L, 122 deg.C, CHEBI:48107, Colorless Yellow, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -42 deg.C, 11349, 63.012
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
1 | |
7697-37-2 | |
122°C | |
Liquid | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 |
Yellow | |
-42°C | |
HNO3 | |
MFCD00011349 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Nitric acid SG 1.42 (70%)
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Nitric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title