missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Honeywell Fluka™
Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone,≥65%
£36.75 - £281.12
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15633090
|
Honeywell-Fluka
30709-1L-GL |
1 L | Glass Bottle |
£36.75
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15623090
|
Honeywell-Fluka
30709-1L |
1 L | Plastic Bottle |
£36.75
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15653090
|
Honeywell-Fluka
30709-2.5L-GL |
2.5 L | Glass Bottle |
£87.86
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15643090
|
Honeywell-Fluka
30709-2.5L |
2.5 L | Plastic Bottle |
£83.80
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15683090
|
Honeywell-Fluka
30709-4X2.5L-GL |
4 x 2.5 L | Glass Bottle |
£281.12
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15673090
|
Honeywell-Fluka
30709-4X2.5L |
4 x 2.5 L | Poly Bottle |
£281.12
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15663090
|
Honeywell-Fluka
30709-40KG |
40 kg | Metal Drum | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
7697-37-2 | |
≤0.0005% (as SO4) | |
HNO3 | |
2031 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01g/mol | |
puriss. p.a. |
1.37 to 1.41g/mL at 20°C | |
8mmHg (20°C) | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
≥65% | |
Nitric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
H272-H290-H314
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Nitric acid
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : nitric acid