Learn More
Thermo Scientific™ Nitrocellulose Membranes, 0.45 μm
Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods).
Brand: Thermo Scientific™ 77010
289.56 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
Code : PN
Additional Details : CAS Number : 9004-70-0 Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods). The 0.45-μm pore size is well-suited for a wide range of western blotting conditions and ideal for the transfer of most proteins (>20 kDa) and nucleic acids (>300 bp).
Features include:
• 100% pure nitrocellulose membrane
• Convenient—save time with pre-cut sheets or choose the roll format for the flexibility to cut to the ideal size
• Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• High-binding affinity—provides excellent protein binding, blocks easily, and does not require pre-wetting with alcohol
• Low background: produce high-quality data with low background signal in both chemiluminescent and fluorescent western blotting
Specifications
12 cm | |
Pierce Nitrocellulose Membrane, 0.45 μm, 12 x 8 cm | |
25 Pre-cut Membranes | |
Nitrocellulose | |
Store membranes flat at ambient temperature, away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. Keep membranes out of direct sunlight. Sunlight may cause the nitrocellulose to dry and become brittle. Note: Nitrocellulose membrane filters are highly flammable. Keep away from heat and open flame. Flash point is approximately 2°C. Membranes can be sterilized by steam-autoclaving at 121°C. | |
12 x 8 cm |
8 cm | |
∼80 μg/cm2 of protein | |
Sheet | |
0.45 μm | |
Room Temperature | |
25 Mini-gel Blots |
Safety and Handling
- Nitrocellulose Membranes, 0.45
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
- MIXTURE LIST-Contains : Nitrocellulose