missing translation for 'onlineSavingsMsg'
Learn More
Nitronium tetrafluoroborate, 0.3 to 0.5M soln. in sulfolane, Thermo Scientific Chemicals Product Code.: 15468765

Nitronium tetrafluoroborate, 0.3 to 0.5M soln. in sulfolane, Thermo Scientific Chemicals

Product Code. 15468765
25 mL
Click to view available options
Quantity:
25 mL
200 mL
This item is not returnable. View return policy

Product Code. 15468765

Brand: Thermo Scientific Alfa Aesar H34069.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
MDL Number MFCD00011432
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolaneShow MoreShow Less
PubChem CID 11073463
IUPAC Name nitrous acid; tetrafluoroboranuide
SMILES [O-][NH+]=O.F[B-](F)(F)F

Especificaciones

Quantity 25 mL
Linear Formula NO2BF4
UN Number UN3265
Formula Weight 132.81
Concentration or Composition (by Analyte or Components) 0.3 to 0.5M soln. in sulfolane
Sensitivity Moisture sensitive
Density 1.268
Chemical Name or Material Nitronium tetrafluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443474 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberH34069.AH,H34069.AC
Lot NumberA0439993 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog NumberH34069.AH,H34069.AC
Lot Number10172694 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34069.AH,H34069.AC

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Nitronium tetrafluoroborate, 0.3 to 0.5M soln. in sulfolane, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado