Learn More
Nitronium tetrafluoroborate, 0.5M solution in sulfolane, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 175130250
74.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Nitronium tetrafluoroborate | |
Colorless through Yellow | |
5.76, 94.97 | |
25 mL | |
NO2BF4 | |
01,742; 04,358; 05,476; 08,361; 09,324; 06,520; 07,324; 09,414; 12,435; 13,274; 14,282; 17,314 | |
Solubility in water: reacts | |
[O-][NH+]=O.F[B-](F)(F)F | |
132.81 | |
132.81 | |
1.268 |
13826-86-3, 126-33-0 | |
5.03, 94.24 | |
Solution | |
BF4NO2 | |
MFCD00011432 | |
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
nitronium;tetrafluoroborate | |
11073463 | |
Glass bottle | |
1.2680g/mL |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Nitronium tetrafluoroborate, 0.5 M solution in sulfolane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 1B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467576 | Certificate of Analysis | 18/12/2024 | 175130250,175132000 |
A0413787 | Certificate of Analysis | 13/12/2022 | 175130250,175132000 |
A0320632 | Certificate of Analysis | 13/12/2022 | 175130250,175132000 |
A0443474 | Certificate of Analysis | 01/08/2022 | 175130250,175132000 |
A0439993 | Certificate of Analysis | 07/04/2022 | 175130250,175132000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only