missing translation for 'onlineSavingsMsg'
Learn More

Nitronium tetrafluoroborate, 0.5M solution in sulfolane, Thermo Scientific Chemicals

£75.07 - £502.50

Specifications

Chemical Name or Material Nitronium tetrafluoroborate
CAS 13826-86-3, 126-33-0
Color Colorless through Yellow
CAS Min % 5.03, 94.24
CAS Max % 5.76, 94.97
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10092380
View Documents
Thermo Scientific Acros
175130250
25 mL Glass bottle
£75.07
25mL
Please to purchase this item. Need a web account? Register with us today!
10732221
View Documents
Thermo Scientific Acros
175132000
200 mL Glass bottle
£502.50
200mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nitronium tetrafluoroborate
Colorless through Yellow
5.76, 94.97
BF4NO2
MFCD00011432
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
XYRQURYIJKJVSF-UHFFFAOYSA-N
nitronium;tetrafluoroborate
11073463
Glass bottle
1.2680g/mL
13826-86-3, 126-33-0
5.03, 94.24
Solution
NO2BF4
01,742; 04,358; 05,476; 08,361; 09,324; 06,520; 07,324; 09,414; 12,435; 13,274; 14,282; 17,314
Solubility in water: reacts
[O-][NH+]=O.F[B-](F)(F)F
132.81
132.81
1.268
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 237-533-5

missing translation for 'rtecsNumber' : QX8620000

missing translation for 'tsca' : TSCA

Product Identifier
  • Nitronium tetrafluoroborate, 0.5 M solution in sulfolane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 1B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only