Learn More
Nitrosonium hexafluorophosphate, 96%, Thermo Scientific Chemicals
£84.02 - £303.51
Chemical Identifiers
CAS | 16921-91-8 |
---|---|
Molecular Formula | F6NOP |
Molecular Weight (g/mol) | 174.97 |
MDL Number | MFCD00040324 |
InChI Key | SAKPNYRUBHJGAR-UHFFFAOYSA-N |
Synonym | nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosoniumShow More |
PubChem CID | 10910083 |
IUPAC Name | azanylidyneoxidanium;hexafluorophosphate |
SMILES | N#[O+].F[P-](F)(F)(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322248
|
Thermo Scientific Alfa Aesar
013149.06 |
5 g |
£84.02
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312248
|
Thermo Scientific Alfa Aesar
013149.14 |
25 g |
£303.51
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nitrosonium hexafluorophosphate is used as initiator in the polymerization reactions of 1,2-epoxides such as styrene oxide, cyclohexene oxide and 1,2-epoxybutane. Further, it is involved in the polymerization of styrene, n-butyl vinyl ether and tetrahydrofuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16921-91-8 | |
174.97 | |
SAKPNYRUBHJGAR-UHFFFAOYSA-N | |
10910083 | |
N#[O+].F[P-](F)(F)(F)(F)F |
F6NOP | |
MFCD00040324 | |
nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosoniumShow More | |
azanylidyneoxidanium;hexafluorophosphate |
Specifications
16921-91-8 | |
F6NOP | |
MFCD00040324 | |
nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium | |
SAKPNYRUBHJGAR-UHFFFAOYSA-N | |
azanylidyneoxidanium;hexafluorophosphate | |
10910083 | |
96% | |
Moisture sensitive |
Crystalline | |
NOPF6 | |
UN3260 | |
Soluble in acetonitrile. | |
N#[O+].F[P-](F)(F)(F)(F)F | |
174.97 | |
174.96 | |
Odorless | |
Nitrosonium hexafluorophosphate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 240-980-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.