Learn More
Invitrogen™ No-Stain™ Protein Labeling Reagent
A fast, easy to use, covalent protein labeling reagent, applied to a membrane after gel transfer that provides sensitive, linear detection of protein for total protein normalization of western blotting data. The Invitrogen No-Stain Reagent can also be used as a fast, sensitive gel stain for visualization of proteins in a gel.
Brand: Invitrogen™ A44449
90.28 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
View more versions of this product
Description
- Flexible use labeling format—Use with any protein gel of choice to perform total protein labeling of the membrane post-transfer. Or use it as a fast protein stain after electrophoresis of gels you do not intend to transfer.
- Easy to use protocol—Mix and incubate with the transferred membrane. The reaction time is 10 minutes. Gel staining is also a 10 minute reaction.
- Flexible visualization—Use a wide-range of imagers with UV, Green LED or Fluorescence (∼488 mm) light source, including the iBright FL1500 Imaging System, to capture an image.
- Accurate total protein normalization—Designed to provide accurate total protein normalization over a broad linear range for protein detection: 1–80 μg total protein loaded per well.
- Sensitive and stable signal—Protein bands are detected down to 20 ng and the signal is compatible with downstream antibody detection.
- Total protein normalization is a useful method for obtaining accurate, quantitative western blotting data, as housekeeping proteins can often be affected by experimental conditions.
Specifications
Protein Labeling Reagent | |
The No-Stain Protein Labeling Reagent forms covalent bonds with the lysine amino acid side chains on all proteins | |
A standard kit will label 40 mini gels or 40 mini gel-sized membranes | |
Dry Ice | |
Any Gel Type and Gel Chemistry | |
Invitrogen™ | |
Compatible with all downstream immunodetection steps (blocking, antibody binding, chemiluminescent or fluorescence based detection |
• No-Stain activator, 800 μL, store at -20°C • No-Stain derivatizer, 800 μL, store at -20°C • No-Stain labeling buffer (20X), 2 x 20 mL, store at 15°C to 30°C |
|
Compatible with wide-range of imagers with UV or Fluorescence light sources, for example, the iBright imager | |
10 minutes | |
Chemiluminescence, Fluorescence | |
PVDF, Nitrocellulose | |
40 reactions |
Safety and Handling
- 40RXN No-Stain Protein Labelling Reagent
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol resistant foam to extinguish.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Mandelonitrile, DMSO
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.