missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ Nonwoven Polyester Lab-Tips™ Swabs

Nonwoven Polyester Swabs are available with small and long handles.

Brand:  Berkshire™ LTN70F.20

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 17208650

  • £565.00 / Pack of 2000
Estimated Shipment: 24-09-2024
to see stock.
Add to basket

Description

Description

  • Double layer, hydroentangled polyester over flexible paddle
  • Soft and non abrasive
  • Thermally bonded head
  • Packaged in an ISO class 4 (Class 10) cleanroom
Specifications

Specifications

RoHS 2002/05 and REACH
2.75 in.
Swab
7cm
5.9cm
1.1cm
0.14 in.
Polypropylene
Non-woven Polyester
0.086 in.
Cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials
0.22cm
2.32 in.
0.433 in.
7mm
Non-contaminating
Flexible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers