missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amyloid beta 42 Human ELISA Kit

Quantifies natural and recombinant Aβ42 selectively in tissue culture medium, tissue homogenate, cerebrospinal fluid, and other samples

Brand:  Invitrogen™ KHB3442

1026.00 GBP valid until 2025-12-31
Use promo code "24347" to get your promotional price.


 View more versions of this product

Product Code. 10561275

  • £1124.00 / 192 tests
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: Pre-coated plate, standard, detector antibody, anti-IgG-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
This item is not returnable. View return policy

Description

Description

The Human Amyloid Beta 42 (Hu Abeta 42) ELISA quantitates Hu Abeta 42 in human tissue culture medium, tissue homogenate, and cerebrospinal fluid (CSF). The assay will exclusively recognize both natural and recombinant Hu Abeta 42. Principle of the method The Human Abeta 42 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Beta-amyloid is a 40–43 amino acid peptide cleaved from amyloid precursor protein, and a putative gamma-secretase. Amyloid beta (1-40) together with Amyloid beta (1-42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the amyloid beta (40) and amyloid beta (42) alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

P05067
<10 pg/mL
Biotin
Cerebrospinal Fluid, Supernatant, Tissue Homogenates
Colorimetric Microplate Reader
AAA,ABETA,ABPP,AD1I,CTFgamma,CVAP,PN-II,PN2,APP
4.3%
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Aβ 42
RUO
2°C to 8°C
1 hr. 20 min.
15.6-1000 pg/mL
15.6 to 1000pg/mL
ELISA Kit
Human
351
4 hr.
3.6%
HRP
2 x 96 Tests
Cerebrospinal Fluid,50 μL; Supernatant, 50 μL; Tissue Homogenate,50 μL
Human
4 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.