missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Protein G for Immunoprecipitation

Product Code. p-4924232
Click to view available options
Quantity:
1 mL
5 mL
50 mL
Unit Size:
5mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10446293

Brand: Invitrogen™ 10003D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dynabeads Protein G beads are uniform, 2.8-μm superparamagnetic beads with recombinant Protein G (∼17 kDa) covalently coupled to the surface.

Dynabeads Protein G beads are uniform, 2.8-μm superparamagnetic beads with recombinant Protein G (∼17 kDa) covalently coupled to the surface. Dynabeads Protein G beads provide a great alternative to Sepharose resin or agarose resin for immunoprecipitation (IP) combined with your own antibody, and both manual and automated protocols are available.

Benefits of Dynabeads Protein G beads include:
Fast and easy—IP in less than 40 minutes with no columns, centrifugations, or time-consuming pre-clearing required
High performance—high target protein yield with low antibody consumption
Low background—very low non-specific binding with a high signal-to-noise ratio
High reproducibility—uniform beads ensure the most consistent results
Automation ready—automation protocols available with the KingFisher Sample Purification instruments

Gentle separation causes minimal physical stress to proteins
The magnetic separation technology utilized by Dynabeads Protein G is rapid and gentle, causing minimal physical stress to target proteins. This permits the isolation and concentration of labile composites that might otherwise dissociate or be damaged by proteases during long incubation times. Native protein conformation and large protein complexes are preserved.

Binding strength and antibody binding capacity
Dynabeads Protein G beads allow for the isolation of most mammalian immunoglobulins (Ig). The amount of Ig captured depends on the concentration of Ig in the starting sample and on the type and source of the Ig. 100 μL of Dynabeads Protein G will isolate approximately 25–30 μg human IgG from a sample containing 20–200 μg IgG/mL. Predominantly Fc-binding allows optimal Ig orientation.

Dynabeads magnetic beads are not porous in nature such as Sepharose or agarose resins. This means that all antibodies bind to the smooth outer surface of the beads and thus are available to bind the target protein. This translates to a low antibody consumption while obtaining a high yield of the target protein. The smooth bead surface is also responsible for the low non-specific binding that Dynabeads magnetic beads are known for.

Dynabeads immunoprecipitation protocol is fast and easy
The manual IP protocol is simple and can be performed in less than 40 minutes with 3 easy bind-incubate-elute steps. The procedure does not require any pre-clearing due to the low non-specific binding and uses very little antibody while still obtaining a high IP yield, thus saving both time and cost per IP.

The antibody-coated beads can be used in a variety of IP applications such as Co-IP, chromatin IP (ChIP/ChIP sequencing), RNA IP (RIP, RIP sequencing), small-scale IgG purification, and protein purification. The isolated pure target protein can be used in downstream western blot, mass spectrometry analysis, sequencing, etc.

Automation-ready for use with KingFisher purification systems
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems or liquid handlers.

Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them the ideal choice for commercial supply. If you are in the process of customizing Dynabeads Protein G beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.

Learn more about Dynabeads magnetic beads and KingFisher Sample Purification Systems:
• Dynabeads Protein G beads are also available as a 'ready-to-go' kit with buffers included (Cat. No. 10007D)
• See our immunoprecipitation selection table, data, and references
• Dynabeads magnetic beads selection guide
• See magnets for Dynabeads magnetic beads separations
• KingFisher automation protocols
• Watch a video about the KingFisher Flex instrument

*Sepharose is a trademark of GE Healthcare Bio-Sciences AB.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 30 mg/mL
Description Recombinant Protein G covalently bound to Dynabeads
Material Polystyrene
Content And Storage Dynabeads Protein G are supplied in PBS, pH 7.4 w/0.01% Tween-20, 0.09% NaN3 Store at 2–8°C.

Diameter (Metric) 2.8 μm
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput Compatibility High-throughput Compatible
Shipping Condition Ambient temperature
Surface Functionality Protein G
Product Line Dynabeads
Quantity 1 mL
For Use With (Application) Immunoprecipitation
Target Antibodies
Type Superparamagnetic Beads
Ligand Type Protein G
Sufficient For 20 Tests
Format Beads in suspension
Purity or Quality Grade For Research Use Only
Certifications/Compliance ISO9001 and ISO13485
Uniformity Monosized 2.8 mm (CV <5%)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.