missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ Transfer Stack, nitrocellulose, mini
GREENER_CHOICE

iBlot Transfer Stacks are used to transfer proteins

Brand:  Invitrogen™ IB301032


 View more versions of this product

Product Code. 12219319

  • £304.00 / Pack of 3
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: One box of iBlot™ Transfer Stacks containing 10 stacks.
This item is not returnable. View return policy

Description

Description

The iBlotTransfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlotTransfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.

Nitrocellulose membrane

  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Bind proteins due to hydrophobic and electrostatic interactions
  • Pore size: 0.2μm
  • Protein binding capacity: 209μg/cm2

 

Transfer stack sizes

  • The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
  • The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)

 

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 cm
Transfer Stack
0.2 μm
Mini, 8 cm x 8 cm
30 Blots
8 cm
Nitrocellulose
30 sets of transfer stacks in 3 boxes. Store at room temperature.
iBlot
Sustainable disposal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • iBlot Transfer Stack, nitrocellulose, mini

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)-1,3-propanediol, Ethylene glycol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.