missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ Transfer Stack, PVDF, mini
GREENER_CHOICE

Used to transfer proteins with the iBlot Gel Transfer Device

Brand:  Invitrogen™ IB401032

318.25 GBP valid until 2025-12-31
Use promo code "24363" to get your promotional price.


 View more versions of this product

Product Code. 11330982

  • £356.00 / Pack of 3
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: One box of iBlot Transfer Stacks containing 10 stacks
This item is not returnable. View return policy

Description

Description

The iBlot Transfer Stacks are used to transfer proteins using the iBlot Gel Transfer Device. The iBlot Transfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlot Transfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.

PVDF membrane

  • 0.2μm pore size, low fluorescence
  • Has higher binding capacity than nitrocellulose
  • Preactivated and ready for use without any pretreatment with alcohols
  • Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Bind proteins due to hydrophobic interactions
  • Protein binding capacity: 240μg/cm2

Transfer stack sizes

  • The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
  • The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 cm
3 x 10 sets
PVDF
30 sets of transfer stacks in 3 boxes. Store at room temperature.
iBlot
Sustainable disposal
8 cm
Transfer Stack
0.2 μm
Mini, 8 cm x 8 cm
30 Blots
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ iBlot™ Transfer Stack, PVDF, mini > Mini, 3 x 10 sets

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.