missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ni-NTA Agarose
Nickel-charged affinity resin that can be used to purify recombinant proteins containing a polyhistidine (6xHis) sequence
£332.00 - £2120.00
Specifications
Content And Storage | The Ni-NTA resin is pre-charged and able to bind up to 50 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 30% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. Ni-NTA resin is guaranteed stable for 6 months when properly stored. |
---|---|
Stationary Phase | Ni-NTA |
Type | Agarose |
Form | Suspension |
Column Type | Affinity Column |
Includes: The Ni-NTA resin is pre-charged and able to bind up to 50mg of recombinant protein per 1mL of resin. It is provided as a 50% slurry in 30% ethanol. The resin will appear blue in color when charged with Ni2+
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11520656
|
Invitrogen™
R90101 |
10 mL |
£332.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11540656
|
Invitrogen™
R90115 |
25 mL |
£566.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11530656
|
Invitrogen™
R90110 |
100 mL |
£2120.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Ni-NTA Agarose is a nickel-charged affinity resin that can be used to purify recombinant proteins containing a polyhistidine (6xHis) sequence
- Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine
- One-step purification can be performed under both native and denaturing conditions
- Ni-NTA uses the chelating ligand nitrilotriacetic acid (NTA) coupled to a cross-linked 6% agarose resin that is suitable for use in batch and gravity flow applications
- Guaranteed stable for 6 months when properly stored at 4°C
Specifications
The Ni-NTA resin is pre-charged and able to bind up to 50 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 30% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. Ni-NTA resin is guaranteed stable for 6 months when properly stored. | |
Agarose | |
Affinity Column |
Ni-NTA | |
Suspension |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Ni-NTA Agarose