missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cell Lysis Buffer II

Cell Lysis Buffer II is a high-quality, ready-to-use lysis buffer suitable for the preparation of cell extracts for ELISA, western blotting, and antibody bead immunoassays (Luminex) applications.

Brand:  Invitrogen™ FNN0021

122.55 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10069262

  • £132.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Cell Lysis Buffer II is a high-quality, ready-to-use lysis buffer suitable for the preparation of cell extracts for ELISA, western blotting, and antibody bead immunoassays (Luminex) applications. The buffer is used for total protein extraction and utilizes detergent-based lysis, eliminating the need for mechanical cell disruption. The formulation helps retain the protein structure and function needed for enzyme assays or immunoassays.

Buffer formulation: 50 mM Tris, pH 7.4, 250 mM NaCl, 5 mM EDTA, 50 mM NaF, 1 mM Na3VO4, 1% detergent, 0.02% NaN3

This cell lysis buffer does not contain protease inhibitors and should be supplemented with 1 mM PMSF and protease inhibitor cocktail (Cat. No. A32955) just prior to use.

Order Info

Shipping condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
Cell Lysis Buffer
100 mL
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number441776-000 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog Number
Lot Number445856-000 Certificate TypeCertificate of Analysis Date01/04/2025 Catalog Number
Lot Number437427-000 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number
Lot Number437427-000 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberFNN0021
Lot Number431739-000 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberFNN0021
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Cell Lysis Buffer II > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.