Learn More
Invitrogen™ PVDF/Filter Paper Sandwich, 0.2 μm, 8.3 x 7.3 cm
Pre-cut and pre-assembled—ready for protein blotting/transfer
Brand: Invitrogen™ LC2002
169.20 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
Code : 50
Additional Details : Weight : 0.11340kg
Description
Twenty membrane/filter paper sandwiches are provided, for 20 transfers. A protein's properties (i.e., charge, hydrophobicity, etc.) affect its ability to bind to membrane surfaces
Applications Western transfer, protein sequencing, amino acid analysis, solid phase assay systems
Dimensions 8.3 x 7.3cm
Pore Size 0.2μm
Binding capacity 50 to 150 μg/cm2 for large, globular proteins; >150μg/cm2 for smaller peptides.
Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
8.3 cm | |
Sandwich | |
0.2 μm | |
Room Temperature | |
0.8 mm |
7.3 cm | |
PVDF | |
Store at room temperature. | |
7.3 cm x 8.3 cm | |
Novex |
For Research Use Only. Not for use in diagnostic procedures.